ENAH30368269 Enamine

1-(butylsulfanyl)-3-(chloromethyl)benzene

  • MDL number MFCD31719411
  • Cas Number : 2253640-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730798

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730798
Availability details for
Quantity
  • Availability
  • Add to Cart