ENAH3036826C Enamine

tert-butyl (2R)-2-aminohexanoate hydrochloride

  • MDL number MFCD31719412
  • Cas Number : 158741-12-9
  • Molecular Weight : 223.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (R)-2-aminohexanoate hydrochloride
Inchi Code 1S/C10H21NO2.ClH/c1-5-6-7-8(11)9(12)13-10(2,3)4;/h8H,5-7,11H2,1-4H3;1H/t8-;/m1./s1
Inchi Key YWUGPFDZLBSQBJ-DDWIOCJRSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6730814

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730814
Availability details for
Quantity
  • Availability
  • Add to Cart