ENAH3036826D Enamine

tert-butyl (2S)-2-aminohexanoate hydrochloride

  • MDL number MFCD31745853
  • Cas Number : 119483-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730815

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730815
Availability details for
Quantity
  • Availability
  • Add to Cart