ENAH3036826E Enamine

1-bromo-4-(2,2-difluorocyclopropyl)benzene

  • MDL number MFCD22548234
  • Cas Number : 1275621-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730818

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730818
Availability details for
Quantity
  • Availability
  • Add to Cart