ENAH30368272 Enamine

2-(2,6-dimethoxybenzoyl)furan

  • MDL number MFCD16513432
  • Cas Number : 1281642-54-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730861

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730861
Availability details for
Quantity
  • Availability
  • Add to Cart