ENAH30368277 Enamine

methyl 1-oxa-8-azaspiro[4.5]dec-3-ene-3-carboxylate hydrochloride

  • MDL number MFCD31719642
  • Cas Number : 2253630-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730929

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730929
Availability details for
Quantity
  • Availability
  • Add to Cart