ENAH3036827C Enamine

5-(chloromethyl)-3-ethyl-1,2,4-thiadiazole

  • MDL number MFCD20638137
  • Cas Number : 1312601-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6730943

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6730943
Availability details for
Quantity
  • Availability
  • Add to Cart