ENAH3036827D Enamine

(2S)-1-(3-fluoro-5-methylbenzoyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD26996265
  • Cas Number : 1608820-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6731034

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6731034
Availability details for
Quantity
  • Availability
  • Add to Cart