ENAH3036827E Enamine

3-bromo-4-ethoxy-2-oxo-2H-pyran-6-carboxylic acid

  • MDL number MFCD31719645
  • Cas Number : 2253640-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6731054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6731054
Availability details for
Quantity
  • Availability
  • Add to Cart