ENAH30368299 Enamine

magnesium(2+) prop-2-ene-1-sulfinate chloride

  • MDL number MFCD31719646

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data ClMg
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6734384

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6734384
Availability details for
Quantity
  • Availability
  • Add to Cart