ENAH3036829B Enamine

7,7-difluoro-1-azaspiro[3.5]nonane hydrochloride

  • MDL number MFCD31719504
  • Cas Number : 2253633-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6526746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6526746
Availability details for
Quantity
  • Availability
  • Add to Cart