ENAH3036829C Enamine

5-bromo-1-chloro-3-(chloromethyl)-2-fluorobenzene

  • MDL number MFCD26686137
  • Cas Number : 1534467-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6526748

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6526748
Availability details for
Quantity
  • Availability
  • Add to Cart