ENAH303682A0 Enamine

5-tert-butylpyridine-2-sulfonyl fluoride

  • MDL number MFCD31719509
  • Cas Number : 2253639-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1582581

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1582581
Availability details for
Quantity
  • Availability
  • Add to Cart