ENAH303682A1 Enamine

1-phenyl-1H-pyrazole-5-sulfonyl fluoride

  • MDL number MFCD31719510
  • Cas Number : 2253630-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1584369

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1584369
Availability details for
Quantity
  • Availability
  • Add to Cart