ENAH303682A9 Enamine

ethyl 2-fluorocyclopent-1-ene-1-carboxylate

  • MDL number MFCD28385643
  • Cas Number : 173277-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6526751

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6526751
Availability details for
Quantity
  • Availability
  • Add to Cart