ENAH303682B4 Enamine

tert-butyl N-[(1-oxo-2,3-dihydro-1H-inden-5-yl)methyl]carbamate

  • MDL number MFCD31736492
  • Cas Number : 954134-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1588692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1588692
Availability details for
Quantity
  • Availability
  • Add to Cart