ENAH303682B7 Enamine

4-[(1H-pyrazol-5-yl)methyl]piperidine hydrochloride

  • MDL number MFCD31719512

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1589182

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1589182
Availability details for
Quantity
  • Availability
  • Add to Cart