ENAH303682C1 Enamine

5-ethylnonan-3-one

  • MDL number MFCD16073757
  • Cas Number : 6137-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6526831

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6526831
Availability details for
Quantity
  • Availability
  • Add to Cart