ENAH303682C4 Enamine

2-bromo-1-methyl-1H,4H,5H,6H,7H-pyrrolo[3,2-c]pyridin-4-one

  • MDL number MFCD31719541
  • Cas Number : 1531541-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6526832

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6526832
Availability details for
Quantity
  • Availability
  • Add to Cart