ENAH303682D2 Enamine

tert-butyl (5E)-4,4-difluoro-5-(2-methoxy-2-oxoethylidene)azepane-1-carboxylate

  • MDL number MFCD31719543
  • Cas Number : 2253644-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6554411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6554411
Availability details for
Quantity
  • Availability
  • Add to Cart