ENAH303682D7 Enamine

4-methylpentane-1,2-diamine dihydrochloride

  • MDL number MFCD31745852
  • Cas Number : 183196-89-6
  • Molecular Weight : 189.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-methylpentane-1,2-diamine dihydrochloride
Inchi Code 1S/C6H16N2.2ClH/c1-5(2)3-6(8)4-7;;/h5-6H,3-4,7-8H2,1-2H3;2*1H
Inchi Key NCDNAUBCSHFYIK-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6632916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6632916
Availability details for
Quantity
  • Availability
  • Add to Cart