ENAH303682DB Enamine

4-(1-hydrazinylethyl)benzoic acid hydrochloride

  • MDL number MFCD31719544
  • Cas Number : 2253630-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6720397

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6720397
Availability details for
Quantity
  • Availability
  • Add to Cart