ENAH303682DF Enamine

6-amino-5-methyl-1,2-dihydropyridin-2-one hydrobromide

  • MDL number MFCD31719533
  • Cas Number : 2253638-61-6
  • Molecular Weight : 205.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Inchi Code 1S/C6H8N2O.BrH/c1-4-2-3-5(9)8-6(4)7;/h2-3H,1H3,(H3,7,8,9);1H
Inchi Key CWWKKQPUZNLBJO-UHFFFAOYSA-N

Description

Purity 95
Salt Data Br
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6728180

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6728180
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart