ENAH303682E6 Enamine

1-[1-(trifluoromethyl)cyclohexyl]methanamine hydrochloride

  • MDL number MFCD31719546
  • Cas Number : 2253631-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6728973

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6728973
Availability details for
Quantity
  • Availability
  • Add to Cart