ENAH303682E7 Enamine

2-(2-aminophenoxy)ethan-1-ol hydrochloride

  • MDL number MFCD31736657
  • Cas Number : 1050652-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6728975

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6728975
Availability details for
Quantity
  • Availability
  • Add to Cart