ENAH303682E9 Enamine

1-(bromomethyl)-2-(trifluoromethyl)cyclohexane

  • MDL number MFCD26921213
  • Cas Number : 1567124-09-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1590077

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1590077
Availability details for
Quantity
  • Availability
  • Add to Cart