ENAH303682EE Enamine

7-bromo-1,3-dihydro-2lambda6,1-benzothiazole-2,2-dione

  • MDL number MFCD31718518
  • Cas Number : 2167917-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1212454

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1212454
Availability details for
Quantity
  • Availability
  • Add to Cart