ENAH303682F2 Enamine

1-(oxetan-3-yl)ethan-1-amine

  • MDL number MFCD26406835
  • Cas Number : 1544892-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1215816

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P260,P261,P264,P271,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P363,P370+P378,P403+P233,P403+P235,P405,P501
hcodes C,Xi
MSDS https://www.enaminestore.com/catalog/EN300-1215816
pictograms GHS02,GHS05,GHS07
hstatements H226,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart