ENAH303682F6 Enamine

2,2-difluoro-1-phenylpropan-1-one

  • MDL number MFCD18811068
  • Cas Number : 703-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1221127

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1221127
Availability details for
Quantity
  • Availability
  • Add to Cart