ENAH303682F7 Enamine

1-methyl-3-(pyridin-2-yl)-1H-pyrazol-5-ol

  • MDL number MFCD28129666
  • Cas Number : 1279105-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1221649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1221649
Availability details for
Quantity
  • Availability
  • Add to Cart