ENAH303682F8 Enamine

4-(piperidin-4-yl)morpholine-3,5-dione hydrochloride

  • MDL number MFCD30004147
  • Cas Number : 2098129-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1222500

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1222500
Availability details for
Quantity
  • Availability
  • Add to Cart