ENAH303682FA Enamine

1-(pyridin-4-yl)azetidin-3-amine dihydrochloride

  • MDL number MFCD28388737
  • Cas Number : 2098025-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1222642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1222642
Availability details for
Quantity
  • Availability
  • Add to Cart