ENAH303682FB Enamine

5-phenyl-1,4-oxazepane

  • MDL number MFCD25983785
  • Cas Number : 933705-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1223135

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1223135
Availability details for
Quantity
  • Availability
  • Add to Cart