ENAH303682FD Enamine

[1-(aminomethyl)-3,3-dimethylcyclobutyl]methanol

  • MDL number MFCD21122027
  • Cas Number : 1489162-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1223295

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1223295
Availability details for
Quantity
  • Availability
  • Add to Cart