ENAH30368301 Enamine

1-(oxan-4-yl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD19612056
  • Cas Number : 1342469-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1223447

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1223447
Availability details for
Quantity
  • Availability
  • Add to Cart