ENAH30368307 Enamine

2,3,6-trimethoxybenzaldehyde

  • MDL number MFCD06739499
  • Cas Number : 5556-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1228692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1228692
Availability details for
Quantity
  • Availability
  • Add to Cart