ENAH30368314 Enamine

1-(3,5-dimethyladamantan-1-yl)methanamine hydrochloride

  • Cas Number : 110916-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 258-260
Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1228788

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1228788
Availability details for
Quantity
  • Availability
  • Add to Cart