ENAH30368315 Enamine

1-chloro-2-ethoxy-2-methylpropane

  • MDL number MFCD19234333
  • Cas Number : 854259-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1228846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1228846
Availability details for
Quantity
  • Availability
  • Add to Cart