ENAH3036833C Enamine

2-{[(tert-butoxy)carbonyl]amino}bicyclo[4.1.0]heptane-2-carboxylic acid

  • MDL number MFCD28389389
  • Cas Number : 1875958-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1229260

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1229260
Availability details for
Quantity
  • Availability
  • Add to Cart