ENAH30368348 Enamine

1-cyclopropylbutan-2-one

  • MDL number MFCD02632331
  • Cas Number : 14113-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1230945

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1230945
Availability details for
Quantity
  • Availability
  • Add to Cart