ENAH30368358 Enamine

2-(cyclopropylsulfanyl)aniline

  • MDL number MFCD26832243
  • Cas Number : 1158702-34-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1238756

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1238756
Availability details for
Quantity
  • Availability
  • Add to Cart