ENAH30368359 Enamine

2-(5-ethylthiophen-2-yl)-2-oxoacetic acid

  • MDL number MFCD11179627
  • Cas Number : 62323-41-5
  • Molecular Weight : 184.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5-ethyl-2-thienyl)(oxo)acetic acid
Inchi Code 1S/C8H8O3S/c1-2-5-3-4-6(12-5)7(9)8(10)11/h3-4H,2H2,1H3,(H,10,11)
Inchi Key JRUKIEVNYGYTAA-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1239560

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1239560
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart