ENAH30368365 Enamine

4-chloro-5,6-dihydro-2H-pyran-3-carboxylic acid

  • MDL number MFCD24135402
  • Cas Number : 1541667-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1241465

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1241465
Availability details for
Quantity
  • Availability
  • Add to Cart