ENAH30368366 Enamine

2-chloro-5-ethyl-6-methylpyridine-3-carbonitrile

  • MDL number MFCD22046770
  • Cas Number : 139393-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-64
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1241556

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1241556
Availability details for
Quantity
  • Availability
  • Add to Cart