ENAH30368370 Enamine

3-(prop-2-yn-1-yl)oxolane-3-carboxylic acid

  • MDL number MFCD21921387
  • Cas Number : 1526588-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1245459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1245459
Availability details for
Quantity
  • Availability
  • Add to Cart