ENAH30368382 Enamine

1-[(propan-2-ylsulfanyl)methyl]cyclohexan-1-ol

  • MDL number MFCD21182494
  • Cas Number : 1470518-64-9
  • Molecular Weight : 188.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-[(isopropylsulfanyl)methyl]cyclohexanol
Inchi Code 1S/C10H20OS/c1-9(2)12-8-10(11)6-4-3-5-7-10/h9,11H,3-8H2,1-2H3
Inchi Key MWPPGTUCHPVWDM-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1249607

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1249607
Availability details for
Quantity
  • Availability
  • Add to Cart