ENAH3036838D Enamine

1-methanesulfinyl-3,3-dimethylbutan-2-one

  • MDL number MFCD20378568
  • Cas Number : 64084-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 25-26
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1250939

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1250939
Availability details for
Quantity
  • Availability
  • Add to Cart