ENAH30368393 Enamine

(3-cyclopropyl-4,5-dihydro-1,2-oxazol-5-yl)methanol

  • MDL number MFCD19666760
  • Cas Number : 1343591-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1251926

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1251926
Availability details for
Quantity
  • Availability
  • Add to Cart