ENAH3036839D Enamine

4-chloro-1-(propan-2-yl)piperidine

  • MDL number MFCD18346568
  • Cas Number : 5570-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1253561

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1253561
Availability details for
Quantity
  • Availability
  • Add to Cart