ENAH3036839E Enamine

1-ethylcycloheptan-1-amine

  • MDL number MFCD12067684
  • Cas Number : 67404-88-0
  • Molecular Weight : 141.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-ethylcycloheptanamine
Inchi Code 1S/C9H19N/c1-2-9(10)7-5-3-4-6-8-9/h2-8,10H2,1H3
Inchi Key WJKBJDQVXXTCDN-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1254086

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P260,P261,P264,P271,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P363,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1254086
pictograms GHS02,GHS05,GHS07
hstatements H226,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart